
4-(Aminomethyl)benzoic acid ≥97.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:56-91-7
:
Formula:C₈H₉NO₂
MDL Number:MFCD00010203
:151.16 g/mol
EINECS:200-297-9
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Nonaqueous Titration):min. 97.0 % (calcd.on anh.substance)
- Solubility in hot Water:almost transparency
- Water:max. 5.0 %
Specification Test Results
Frequently Bought Together