
4,4'-Diaminobenzanilide ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Certificaten
Over dit artikel
CAS-nummer:785-30-8
:
Formule:C₁₃H₁₃N₃O
MDL-nummer:MFCD00025361
:227.27 g/mol
Smelten Pt:214…215 °C
EINECS:212-321-5
:GHS07
Specificatie testresultaten
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
Specificatie testresultaten
Frequently Bought Together