Bestelling invoeren
Netherlands
ContactUsLinkComponent
3,5-Dimethoxybenzamide ≥98.0% (by total nitrogen basis)
Irritant
3,5-Dimethoxybenzamide ≥98.0% (by total nitrogen basis)
Catalog # TCIAD1558-5G
Supplier:  TCI
CAS Number:  
undefined
Irritant
3,5-Dimethoxybenzamide ≥98.0% (by total nitrogen basis)
Catalog # TCIAD1558-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Verpakking:
    Glass bottle
  • VPE:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystaline
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    145.0 to 149.0 deg-C

Specification Test Results

About this item

 :
 :181.19 g/mol
Boiling Pt:345 °C (1013 hPa)
Melting Pt:148 °C
EINECS:241-257-0
 :GHS07
CAS Number:17213-58-0
Formula:C₉H₁₁NO₃
MDL Number:MFCD00007987