Order Entry
Netherlands
ContactUsLinkComponent
3,3'-Diaminobenzophenone ≥95.0% (by GC, titration analysis)
Irritant
Health
3,3'-Diaminobenzophenone ≥95.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
Health
3,3'-Diaminobenzophenone ≥95.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange powder to crystal
  • Purity (GC):
    min. 95.0 %
  • Purity (Nonaqueous Titration):
    min. 95.0 %

Specification Test Results

Related Information

Frequently Bought Together