β-Naphthoflavone ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:6051-87-2
:
Formula:C₁₉H₁₂O₂
MDL Number:MFCD00004986
:272.3 g/mol
Boiling Pt:461 °C (1013 hPa)
Melting Pt:163…164 °C
Density:1,11 g/cm³ (20 °C)
EINECS:227-958-4
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:162.0 to 166.0 deg-C
Specification Test Results
Frequently Bought Together