Order Entry
Netherlands
ContactUsLinkComponent
3,5-Dichlorobenzenesulphonamide ≥98.0% (by HPLC)
 
3,5-Dichlorobenzenesulphonamide ≥98.0% (by HPLC)
  TCIAD3502-5G
 :  TCI
 :  
undefined
 
3,5-Dichlorobenzenesulphonamide ≥98.0% (by HPLC)
  TCIAD3502-5G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 98.0 area%

 

 

 :
 :226.08 g/mol
 :GHS07
 :19797-32-1
 :C₆H₅Cl₂NO₂S
 :MFCD00117161