

2-Chloroethanesulphonyl chloride ≥95.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1622-32-8
:
Formula:C₂H₄Cl₂O₂S
MDL Number:MFCD00007461
:163.02 g/mol
Boiling Pt:203°C
Melting Pt:3 °C
Density:1,555 g/cm³ (20 °C)
Flash Pt:152°C
Storage Temperature:Refrigerator
EINECS:216-594-1
UN:2927
ADR:6.1,II
:GHS05
:GHS06
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 95.0 %
- Purity (Argentmetric Titration):min. 95.0 %
Specification Test Results
Frequently Bought Together