
5-Chloro-2,4-dimethoxyaniline ≥97.0% (by GC, titration analysis)
Catalog # TCIAC1226-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Amber to Dark purple powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
- Melting point:90.0 to 93.0 deg-C
Specification Test Results
About this item
:
:187.63 g/mol
Boiling Pt:287 °C (1013 hPa)
Melting Pt:91…92 °C
EINECS:202-586-5
:GHS07
CAS Number:97-50-7
Formula:C₈H₁₀ClNO₂
MDL Number:MFCD00025752