Order Entry
Netherlands
ContactUsLinkComponent
2,3-Dichloro-5-(trifluoromethyl)pyridine ≥95.0% (by GC)
Corrosive
Irritant
Environment
2,3-Dichloro-5-(trifluoromethyl)pyridine ≥95.0% (by GC)
Catalog # TCIAD2214-25G
Supplier:  TCI
CAS Number:  
Corrosive
Irritant
Environment
2,3-Dichloro-5-(trifluoromethyl)pyridine ≥95.0% (by GC)
Catalog # TCIAD2214-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to lump
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
 :215.99 g/mol
Boiling Pt:80°C
Flash Pt:79°C
EINECS:410-340-5
UN:3082
ADR:9,III
 :GHS05
 :GHS07
 :GHS09
CAS Number:69045-84-7
Formula:C₆H₂Cl₂F₃N
MDL Number:MFCD00042243