Order Entry
Netherlands
ContactUsLinkComponent
4,5-Dichloroimidazole ≥97.0% (by GC, titration analysis)
Irritant
4,5-Dichloroimidazole ≥97.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
4,5-Dichloroimidazole ≥97.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Purity (Nonaqueous Titration):
    min. 97.0 %
  • Melting point:
    180.0 to 184.0 deg-C
  • Solubility in Methanol:
    very faint turbidity

Specification Test Results

Frequently Bought Together