Order Entry
Netherlands
ContactUsLinkComponent
4-Bromo-2,5-difluorobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
Corrosive
4-Bromo-2,5-difluorobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
Catalogus # TCIAB2388-1G
Supplier:  TCI
CAS Number:  
undefined
Corrosive
4-Bromo-2,5-difluorobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
Catalogus # TCIAB2388-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Argentmetric Titration):
    min. 98.0 %
  • Melting point:
    39.0 to 43.0 deg-C

Specification Test Results

About this item

 :
 :291.5 g/mol
Boiling Pt:76 °C (0,01 -rr)
Melting Pt:34…36 °C
UN:3261
ADR:8,III
 :GHS05
CAS Number:207974-14-9
Formula:C₆H₂BrClF₂O₂S
MDL Number:MFCD00042181