
2,2-Dibromopropane ≥98.0% (by GC)
Catalog # TCIAD1409-10G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:Colorless to Light red to Green clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
About this item
:
:201.89 g/mol
Boiling Pt:113°C
Melting Pt:–20 °C
Density:1,8149 g/cm³ (0 °C)
EINECS:209-828-9
:GHS07
CAS Number:594-16-1
Formula:H₃CBr₂CCH₃
MDL Number:MFCD00000119