

1,2,3,4-Tetrahydronaphthalene ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:119-64-2
:
Formula:C₁₀H₁₂
MDL Number:MFCD00001733
:132.21 g/mol
Merck Index:14,09221
Boiling Pt:207°C
Melting Pt:–36°C
Density:0,97 g/cm³ (20 °C)
Flash Pt:75°C
Storage Temperature:Ambient
EINECS:204-340-2
UN:3082
ADR:9,III
:GHS07
:GHS09
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together