Order Entry
Netherlands
ContactUsLinkComponent
2-Aminopentane ≥97.0% (by GC)
 
 
2-Aminopentane ≥97.0% (by GC)
  TCIAA1102.25
 :  TCI
 :  
undefined
 
 
2-Aminopentane ≥97.0% (by GC)
  TCIAA1102.25
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

 

 

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 97.0 %

 

 

 :
 :87.17 g/mol
 :91°C
 :-6°C
 :–6°C
 :264-269-8
 :1106
 :3,II
 :GHS02
 :GHS05
 :63493-28-7
 :41444-43-3
 :C₅H₁₃N
 :MFCD00008098