Bestelling invoeren
Netherlands
ContactUsLinkComponent
Bilirubine
Bilirubine
Supplier:  TCI
Certificates
undefined
Bilirubine
Supplier:  TCI
Specification Test Results
  • Appearance:
    Orange to Brown powder to crystal
  • Lambda max.:
    450.0 to 454.0 nm (CHCl3)
  • Molar extinction coefficient:
    59,900 to 61,500 (CHCl3, 450.0 to 454.0 nm)