
4,4'-Dimethoxybenzophenone ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:90-96-0
:
Formula:C₁₅H₁₄O₃
MDL Number:MFCD00008404
:242.27 g/mol
Boiling Pt:180 - 240 °C (13 -rr)
Melting Pt:142…145 °C
Density:1,259 g/cm³ (20 °C)
EINECS:202-028-0
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (GC):min. 99.0 %
- Melting point:144.0 to 147.0 deg-C
- Solubility in hot Toluene:almost transparency
Specification Test Results
Frequently Bought Together