Order Entry
Netherlands
ContactUsLinkComponent
(-)-Quinine hydrochloride dihydrate ≥98.0% (by titrimetric analysis)
Irritant
Health
(-)-Quinine hydrochloride dihydrate ≥98.0% (by titrimetric analysis)
Catalog # TCIAQ0030-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
Health
(-)-Quinine hydrochloride dihydrate ≥98.0% (by titrimetric analysis)
Catalog # TCIAQ0030-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 90.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 % (calcd.on dried substance)
  • Specific rotation [a]20/D:
    -245 to -255 deg (C=2, 0.1mol/l HCl (calcd.on dried substance)
  • Solubility in hot Water:
    almost transparency

Specification Test Results

About this item

 :
 :396.91 g/mol
Merck Index:14,08061
Melting Pt:158…160 °C
Flash Pt:122 °C
EINECS:205-001-1
 :GHS07
 :GHS08
Storage Temperature:Ambient
CAS Number:6119-47-7
Formula:C₂₀H₂₅ClN₂O₂·2H₂O
MDL Number:MFCD00078498