Alizarine ≥95.0% (by HPLC)
Catalog # TCIAD0242-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Orange to Brown powder to crystal
- Purity (HPLC):min. 95.0 area%
- Lambda max.:429.0 to 433.0 nm (MeOH)
- Absorbance:min. 0.400 (20mg/L, MeOH, 429.0 to 433.0 nm)
Specification Test Results
About this item
:
:240.22 g/mol
Merck Index:14,00251
Boiling Pt:430 °C (1013 hPa)
Melting Pt:290 °C
Density:1,54 g/cm³ (20 °C)
EINECS:200-782-5
:GHS07
:GHS08
Storage Temperature:Ambient
CAS Number:72-48-0
Formula:C₁₄H₈O₄
MDL Number:MFCD00001201