Order Entry
Netherlands
ContactUsLinkComponent
2,3-Dimethylheptane ≥99.0% (by GC)
Flammable
Health
2,3-Dimethylheptane ≥99.0% (by GC)
Catalog # TCIAD1203-1ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Health
2,3-Dimethylheptane ≥99.0% (by GC)
Catalog # TCIAD1203-1ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Seq:
    2070

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 99.0 %

Specification Test Results

About this item

 :
 :128.26 g/mol
Boiling Pt:140°C
Melting Pt:–112 °C
Density:0,726 g/cm³ (20 °C)
Flash Pt:26°C
UN:1920
ADR:3,III
 :GHS02
 :GHS08
CAS Number:3074-71-3
Formula:C₉H₂₀
MDL Number:MFCD00048819