α-Hydroxy-o-tolyl-β-D-glucopyranoside ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:138-52-3
:
Formula:C₁₃H₁₈O₇
MDL Number:MFCD00006590
:286.28 g/mol
Merck Index:14,08324
Melting Pt:199…201 °C
Density:1,42 g/cm³ (20 °C)
EINECS:205-331-6
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:199.0 to 202.0 deg-C
- Specific rotation [a]20/D:-60.0 to -64.0 deg (C=3, H2O)
Specification Test Results
Frequently Bought Together