Order Entry
Netherlands
ContactUsLinkComponent
2'-Hydroxypropiophenone ≥97.0% (by GC)
Irritant
2'-Hydroxypropiophenone ≥97.0% (by GC)
Catalog # TCIAH0298-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
2'-Hydroxypropiophenone ≥97.0% (by GC)
Catalog # TCIAH0298-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Yellow to Green clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :150.18 g/mol
Boiling Pt:150°C
Melting Pt:20…22 °C
Density:1,106 g/cm³ (17 °C)
EINECS:210-244-1
 :GHS07
CAS Number:610-99-1
Formula:C₉H₁₀O₂
MDL Number:MFCD00002220