Order Entry
Netherlands
ContactUsLinkComponent
3,4'-Dichloropropiophenone ≥98.0% (by GC, titration analysis)
Corrosive
3,4'-Dichloropropiophenone ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Corrosive
3,4'-Dichloropropiophenone ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Yellow to Orange powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Argentmetric Titration):
    min. 98.0 %
  • Melting point:
    49.0 to 53.0 deg-C
  • Solubility in Acetone:
    almost transparency

Specification Test Results

Frequently Bought Together