
2-Fluoro-5-nitrobenzonitrile ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:17417-09-3
:
Formula:C₇H₃FN₂O₂
MDL Number:MFCD00042299
:166.11 g/mol
Boiling Pt:94°C
Melting Pt:79 °C
UN:3439
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to Yellow to Green powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:71.0 to 75.0 deg-C
Specification Test Results
Frequently Bought Together