6-Amino-m-cresol ≥98.0% (by GC, titration analysis)
: TCI
: 1-Amino-2-hydroxy-4-methylbenzene, 2-Hydroxy-4-methylaniline, Cetilistat Impurity 4, 4-Amino-3-hydroxytoluene, 2-Amino-5-methylphenol
:2835-98-5
:
:C₇H₉NO
:MFCD00007693
:123.15 g/mol
:259 °C (1013 hPa)
:162 °C
:220-620-7
:GHS07
- Environmentally Preferable:
- Appearance:White to Brown powder to crystalline
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Frequently Bought Together