
2,6-Dimethoxy-p-cresol ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:6638-05-7
:
Formula:C₉H₁₂O₃
MDL Number:MFCD00017289
:168.19 g/mol
Boiling Pt:150°C
EINECS:229-641-6
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White or Colorless to Light orange to Yellow powder to lump to clear liquid
- Purity (GC):min. 97.0 %
- Melting point:40.0 to 43.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together