
2,6-Dibromo-p-cresol ≥98.0% (by GC, titration analysis)
TCIAD3487-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:46.0 to 50.0 deg-C
:
:265.93 g/mol
:140°C
:2 °C
:2,22 g/cm³ (20 °C)
:219-404-5
:GHS07
:2432-14-6
:C₇H₆Br₂O
:MFCD00002154