

Diethylene glycol monomethyl ether ≥99.0% (by GC) stabilised
Supplier: TCI
Certificates
About this item
CAS Number:111-77-3
:
Formula:C₅H₁₂O₃
MDL Number:MFCD00002871
:120.15 g/mol
Merck Index:14,06037
Boiling Pt:193°C
Melting Pt:–65 °C
Density:1,025 g/cm³ (20 °C)
Flash Pt:105°C
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
Storage Temperature:Ambient
EINECS:203-906-6
:GHS07
:GHS08
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.0 %
Specification Test Results
Frequently Bought Together