
Tris(pentafluorophenyl)phosphine ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1259-35-4
:
Formula:C₁₈F₁₅P
MDL Number:MFCD00079654
:532.15 g/mol
EINECS:215-021-2
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 95.0 %
- Melting point:114.0 to 118.0 deg-C
Specification Test Results
Frequently Bought Together