Order Entry
Netherlands
ContactUsLinkComponent
 
 
Dodecyltrichlorosilane ≥97.0% (by titrimetric analysis)
  TCIAD1509-25ML
 :  TCI
 :  
undefined
 
Dodecyltrichlorosilane ≥97.0% (by titrimetric analysis)
  TCIAD1509-25ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

 

 

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 96.0 %
  • Purity (Argentmetric Titration):
    min. 97.0 %

 

 

 :
 :303.77 g/mol
 :293 °C (1013 hPa)
 :1,03 g/cm³ (20 °C)
 :165°C
 :224-769-9
 :1771
 :8,II
 :GHS05
 :4484-72-4
 :C₁₂H₂₅Cl₃Si
 :MFCD00000483