cis-3-Nonen-1-ol ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:10340-23-5
:
Formula:C₉H₁₈O
MDL Number:MFCD00010314
Boiling Pt:109°C
Flash Pt:102°C
:142.24 g/mol
EINECS:233-735-2
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together