Order Entry
Netherlands
ContactUsLinkComponent
cis-2-Nonen-1-ol ≥95.0% (by GC)
cis-2-Nonen-1-ol ≥95.0% (by GC)
Catalog # TCIAN0586-10ML
Supplier:  TCI
CAS Number:  
undefined
cis-2-Nonen-1-ol ≥95.0% (by GC)
Catalog # TCIAN0586-10ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • Seq:
    2275

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
 :142.24 g/mol
Boiling Pt:106°C
Flash Pt:101°C
EINECS:255-376-0
CAS Number:41453-56-9
Formula:C₉H₁₈O
MDL Number:MFCD00063210