Order Entry
Netherlands
ContactUsLinkComponent
Phenyl chloroformate ≥98.0% (by GC, titration analysis)
Phenyl chloroformate ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Phenyl chloroformate ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Argentmetric Titration):
    min. 98.0 %

Specification Test Results

Frequently Bought Together