

5-Methyl-1H-tetrazole ≥98.0% (by HPLC, titration analysis)
TCIAM1069-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:145.0 to 148.0 deg-C
- Solubility in Water:almost transparency
:
:84.08 g/mol
:1325
:4.1,II
:GHS02
:GHS07
:4076-36-2
:C₂H₄N₄
:MFCD00129971