
4-Methoxybenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:98-68-0
:
Formula:C₇H₇ClO₃S
MDL Number:MFCD00007446
Boiling Pt:173°C
Melting Pt:40…42 °C
Storage Temperature:Refrigerator
EINECS:202-692-1
UN:3261
ADR:8,II
:206.65 g/mol
:GHS05
Specification Test Results
- Appearance:White to Almost white powder to lump
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together