
4-(Aminomethyl)pyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:3731-53-1
:
Formula:C₆H₈N₂
MDL Number:MFCD00006449
:108.14 g/mol
Boiling Pt:230°C
Melting Pt:–8 °C
Density:1,0688 g/cm³ (20 °C)
Flash Pt:104°C
EINECS:223-092-6
UN:2735
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Colorless to Red to Green clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together