
3-Fluoro-5-(trifluoromethyl)benzaldehyde ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:188815-30-7
:
Formula:C₈H₄F₄O
MDL Number:MFCD00061312
:192.11 g/mol
Flash Pt:71°C
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together