Order Entry
Netherlands
ContactUsLinkComponent
Deoxycholic acid ≥98.0% (by GC, titration analysis)
Irritant
Deoxycholic acid ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
Deoxycholic acid ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    173.0 to 178.0 deg-C
  • Specific rotation [a]20/D:
    +54.0 to +58.0 deg (C=1, EtOH)

Specification Test Results

Related Information

Frequently Bought Together