Order Entry
Netherlands
ContactUsLinkComponent
3,3'-Thiodipropionic acid ≥99.0% (by titrimetric analysis)
Irritant
3,3'-Thiodipropionic acid ≥99.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
Irritant
3,3'-Thiodipropionic acid ≥99.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 99.0 %
  • Melting point:
    130.0 to 134.0 deg-C
  • Solubility in Methanol:
    very faint turbidity

Specification Test Results

Frequently Bought Together