
3-(4-Methoxyphenyl)propionic acid ≥98.0% (by GC, titration analysis)
: TCI
:1929-29-9
:
:C₁₀H₁₂O₃
:MFCD00002777
:180.2 g/mol
:194°C
:95…102 °C
:217-678-0
:GHS07
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:100.0 to 105.0 deg-C
Frequently Bought Together