2-Mercapto-1-methylimidazole ≥98.0% (by HPLC, titration analysis)
TCIAM0868-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:144.0 to 148.0 deg-C
- Solubility in Water:almost transparency
:
:114.17 g/mol
:14,05971
:155 °C (1013 hPa)
:1,36 g/cm³ (20 °C)
:200-482-4
:GHS07
:GHS08
:60-56-0
:C₄H₆N₂S
:MFCD00179321