
2-Chloro-5-(trifluoromethyl)pyridine ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:52334-81-3
:
Formula:C₆H₃ClF₃N
MDL Number:MFCD00042225
:181.54 g/mol
Boiling Pt:147°C
Melting Pt:30…31 °C
Density:1,41 g/cm³ (20 °C)
Flash Pt:65°C
EINECS:257-856-5
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:White or Colorless to Light orange to Yellow powder to lump to clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together