Order Entry
Netherlands
ContactUsLinkComponent
 
 
2-Chloro-5-(trifluoromethyl)benzaldehyde ≥95.0% (by GC)
 :  TCI
 
undefined
 
2-Chloro-5-(trifluoromethyl)benzaldehyde ≥95.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Yellow to Green clear liquid
  • Purity (GC):
    min. 95.0 %

 

Frequently Bought Together