Order Entry
Netherlands
ContactUsLinkComponent
2-Amino-4,6-dichloropyrimidine ≥98.0% (by total nitrogen basis)
Irritant
2-Amino-4,6-dichloropyrimidine ≥98.0% (by total nitrogen basis)
Catalog # TCIAA1110-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
2-Amino-4,6-dichloropyrimidine ≥98.0% (by total nitrogen basis)
Catalog # TCIAA1110-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Brown powder to crystalline
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    222.0 to 226.0 deg-C

Specification Test Results

About this item

 :
 :163.99 g/mol
Melting Pt:216…220 °C
EINECS:200-253-9
 :GHS07
CAS Number:56-05-3
Formula:C₄H₃Cl₂N₃
MDL Number:MFCD00006090