
2,7-Dibromonaphthalene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:58556-75-5
:
Formula:C₁₀H₆Br₂
MDL Number:MFCD00238580
:285.97 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:139.0 to 143.0 deg-C
Specification Test Results
Frequently Bought Together