Order Entry
Netherlands
ContactUsLinkComponent
2,6-Dibromonaphthalene ≥98.0% (by GC)
Irritant
2,6-Dibromonaphthalene ≥98.0% (by GC)
Catalog # TCIAD4154-1G
Supplier:  TCI
CAS Number:  
undefined
Irritant
2,6-Dibromonaphthalene ≥98.0% (by GC)
Catalog # TCIAD4154-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Gray to Brown powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    160.0 to 164.0 deg-C

Specification Test Results

About this item

 :
 :285.97 g/mol
 :GHS07
CAS Number:13720-06-4
Formula:C₁₀H₆Br₂
MDL Number:MFCD01026462