
2,3-Bis(trifluoromethyl)pyridine ≥95.0% (by GC)
Catalog # TCIAB2394-1G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
About this item
:
:215.1 g/mol
Boiling Pt:158°C
Melting Pt:2°C
Flash Pt:70°C
:GHS07
CAS Number:1644-68-4
Formula:C₇H₃F₆N
MDL Number:MFCD03093763