

1-Methyl-3-phenylpiperazine ≥98.0% (by GC, titration analysis)
TCIAM1685-5G
: TCI
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:58.0 to 62.0 deg-C
:
:176.26 g/mol
:431-180-2
:6.1,III
:GHS05
:GHS07
:5271-27-2
:C₁₁H₁₆N₂
:MFCD03411603