Order Entry
Netherlands
ContactUsLinkComponent
4,4'-Dihydroxybenzophenone ≥98.0% (by GC, titration analysis)
Irritant
4,4'-Dihydroxybenzophenone ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
4,4'-Dihydroxybenzophenone ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light red to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    218.0 to 223.0 deg-C
  • Solubility in Methanol:
    almost transparency

Specification Test Results

Related Information

Frequently Bought Together