

1-Hexyl-3-methylimidazolium tetrafluoroborate ≥97.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:244193-50-8
:
Formula:C₁₀H₁₉BF₄N₂
MDL Number:MFCD03095459
:254.08 g/mol
Boiling Pt:? 200 °C (1013 hPa)
Melting Pt:–82 °C
Density:1,14 g/cm³ (20 °C)
Flash Pt:≥ 100 °C
UN:1760
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Appearance:Light yellow to Brown clear liquid
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 97.0 %
Specification Test Results
Related Information
Frequently Bought Together