
2-Fluoro-N-methylaniline ≥98.0% (by GC, titration analysis)
Catalog # TCIAF0449-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
About this item
:
:125.15 g/mol
:GHS07
CAS Number:1978-38-7
Formula:C₇H₈FN
MDL Number:MFCD00028030